5-(Trifluoromethyl)pyridine-2-carboxaldehyde 95%
Lieferant: Thermo Scientific
Achtung
Synonyme: 5-(Trifluoromethyl)picolinaldehyde
H31858.MD H31858.03
H31858.MDEA 156 EUR
H31858.MD H31858.03
5-(Trifluoromethyl)pyridine-2-carboxaldehyde 95%
5-(Trifluoromethyl)pyridine-2-carboxaldehyde
Purity: 95% CAS No.: 31224-82-5 Molecular Formula: C7H4F3NO Formula Weight: 175.11 Storage Temperature: 2°C to 8°C Melting Point: 31-33° MDL No.: MFCD07375388
Formel: C₇H₄F₃NO
Molecular Weight: 175,11 g/mol
Schmelzpunkt: 31…33 °C
MDL: MFCD07375388
CAS-Nummer: 31224-82-5