Dinatriumtetrachloropalladat Hydrat (≥30% Pd) ≥99,999% (Metall-Basis), Premion®
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
Synonyme:
Natriumtetrachloropalladat(II), Natrium-Palladium(II)-chlorid
43945.03
43945.06
43945.03EA
185
EUR
43945.03
43945.06
Dinatriumtetrachloropalladat Hydrat (≥30% Pd) ≥99,999% (Metall-Basis), Premion®
Dinatriumtetrachloropalladat Hydrat
Purity: 99.999%
CAS No.: 13820-53-6
Molecular Formula: Cl4Na2Pd.xH2O (x≈3)
Formula Weight: 294.20(anhy)
Physical Form: Crystalline
Sensitivity: Hygroscopic
Appearance: Red-brown
MDL No.: MFCD00003487
Formel:
Cl₄Na₂Pd·xH₂O Molecular Weight: 294.21 g/mol (anhy) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00003487 CAS-Nummer: 13820-53-6 EINECS: 237-502-6 |