N-Boc-N-ethylglycine 97%
Lieferant: Thermo Scientific
|
Achtung
|
Synonyme:
Boc-N-Ethylglycine, N-Ethylglycine, N-BOC protected, Boc-N-Et-Gly-OH
H32112.03
H32112.06
H32112.03EA
46
EUR
H32112.03
H32112.06
N-Boc-N-ethylglycine 97%
N-Boc-N-ethylglycine
Purity: 97%
CAS No.: 149794-10-5
Molecular Formula: C9H17NO4
Formula Weight: 203.24
Melting Point: 87-88°
MDL No.: MFCD00672508
Formel:
C₉H₁₇NO₄ Molecular Weight: 203,24 g/mol Schmelzpunkt: 87…88 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00672508 CAS-Nummer: 149794-10-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...