2-(tert-Butoxycarbonylamino)-1-ethanol 95%
Lieferant: Thermo Scientific
|
Achtung
|
Synonyme:
N-(2-Hydroxyethyl)carbamic Acid tert-Butyl Ester, 2-(tert-Butoxycarbonyl)ethanolamine, N-(tert-Butoxycarbonyl)ethanolamine, 2-Boc-ethanolamine, tert-Butyl N-(2-Hydroxyethyl)carbamate, 2-(Boc-amino)-1-ethanol, Boc-Glycinol
H32356.06
H32356.14
H32356.06EA
58.6
EUR
H32356.06
H32356.14
2-(tert-Butoxycarbonylamino)-1-ethanol 95%
2-(tert-Butoxycarbonylamino)-1-ethanol
Purity: 95%
CAS No.: 26690-80-2
Molecular Formula: C7H15NO3
Formula Weight: 161.20
Refractive Index: 1.4490
Boiling Point: 92°/0.22mm
MDL No.: MFCD00056657
Formel:
C₇H₁₅NO₃ Molecular Weight: 161,20 g/mol Siedepunkt: 92 °C (0,22 mmHg) Dichte: 1,042 Lagertemperatur: Raumtemperatur |
MDL:
MFCD00056657 CAS-Nummer: 26690-80-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...