(+/-)-2-(Trifluoromethyl)piperazine 97%

Lieferant: Thermo Scientific
Gefahr

Synonyme: 2-(Trifluoromethyl)piperazine

H32524.MD H32524.03
H32524.MDEA 96.9 EUR
H32524.MD H32524.03
(+/-)-2-(Trifluoromethyl)piperazine 97%
(+/-)-2-(Trifluoromethyl)piperazine

Purity: 97% CAS No.: 131922-05-9 Molecular Formula: C5H9F3N2 Formula Weight: 154.13 Melting Point: 94-97° UN No.: UN3259 MDL No.: MFCD07373382

Formel: C₅H₉F₃N₂
Molecular Weight: 154,13 g/mol
Schmelzpunkt: 94…97 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD07373382
CAS-Nummer: 131922-05-9
ADR: 8,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR