2,5-Dimethylthiophene-3-boronic acid 95%

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: (2,5-Dimethylthiophen-3-yl)boronic acid

H32543.MD H32543.03
H32543.MDEA 127 EUR
H32543.MD H32543.03
2,5-Dimethylthiophene-3-boronic acid 95%
2,5-Dimethylthiophene-3-boronic acid

Purity: 95% CAS No.: 162607-23-0 Molecular Formula: C6H9BO2S Formula Weight: 156.01 Storage Temperature: 2°C to 8°C MDL No.: MFCD09837620

Formel: C₆H₉BO₂S
Molecular Weight: 156.01 g/mol
MDL: MFCD09837620
CAS-Nummer: 162607-23-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR