5-Methyl-1H-pyrazol-4-carbaldehyd 97%

Lieferant: Thermo Scientific
Achtung

Synonyme: 4-Formyl-3-methylpyrazole, 3-Methylpyrazole-4-carboxaldehyde

H32547.MD H32547.03
H32547.MDEA 64.6 EUR
H32547.MD H32547.03
5-Methyl-1H-pyrazol-4-carbaldehyd 97%
3-Methyl-1H-pyrazole-4-carbaldehyde

Purity: 97% CAS No.: 112758-40-4 Molecular Formula: C5H6N2O Formula Weight: 110.12 MDL No.: MFCD01313822

Formel: C₅H₆N₂O
Molecular Weight: 110,12 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD01313822
CAS-Nummer: 112758-40-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR