2-Methyl-2-propanyl-N-(diphenylmethylen)glycinat 99%

Lieferant: Thermo Fisher Scientific

Synonyme: N-(Diphenylmethylene)glycine tert-butyl ester, Glycine, N-(diphenylmethylene)-, 1,1-dimethylethyl ester, tert-Butyl 2-((diphenylmethylene)amino)acetate

H32709.03 H32709.06
H32709.03EA 54 EUR
H32709.03 H32709.06
2-Methyl-2-propanyl-N-(diphenylmethylen)glycinat 99%
tert-Butyl-2-((diphenylmethylene)amino)acetate

Purity: 99% CAS No.: 81477-94-3 Molecular Formula: C19H21NO2 Formula Weight: 295.38 Melting Point: 113-115° MDL No.: MFCD00134280

Formel: C₁₉H₂₁NO₂
Molecular Weight: 295.382 g/mol
Schmelzpunkt: 113…115 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00134280
CAS-Nummer: 81477-94-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR