1-(2,6-Difluorophenyl)ethanol 97%
Lieferant: Thermo Scientific
Synonyme: 2,6-Difluoro-alpha-methylbenzyl alcohol
H32771.03 H32771.09
H32771.03EA 40.9 EUR
H32771.03 H32771.09
1-(2,6-Difluorophenyl)ethanol 97%
1-(2,6-Difluorophenyl)ethanol
Purity: 97% CAS No.: 87327-65-9 Molecular Formula: C8H8F2O Formula Weight: 158.15 Refractive Index: 1.4860 MDL No.: MFCD00274321
Formel: C₈H₈F₂O
Molecular Weight: 158,15 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00274321
CAS-Nummer: 87327-65-9