5-Hydroxynicotinonitrile 97%

Lieferant: Thermo Scientific
Gefahr

H33006.MD H33006.03
H33006.MDEA 81.1 EUR
H33006.MD H33006.03
5-Hydroxynicotinonitrile 97%
5-Hydroxynicotinonitrile

Purity: 97% CAS No.: 152803-24-2 Molecular Formula: C6H4N2O Formula Weight: 120.11 Melting Point: 213-217° UN No.: UN3439 MDL No.: MFCD11044307

Formel: C6H4N2O
Molecular Weight: 120,11 g/mol
Schmelzpunkt: 213…217 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD11044307
CAS-Nummer: 152803-24-2
ADR: 6.1,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR