2-Chlor-4,6-diphenyl-1,3,5-triazin 97%
Lieferant: Thermo Scientific
|
Achtung
|
H33175.03
H33175.06
H33175.14
H33175.03EA
29.8
EUR
H33175.03
H33175.06
H33175.14
2-Chlor-4,6-diphenyl-1,3,5-triazin 97%
2-Chloro-4,6-diphenyl-1,3,5-triazine
Purity: 97%
CAS No.: 3842-55-5
Molecular Formula: C15H10ClN3
Formula Weight: 267.72
Melting Point: 138-140°
MDL No.: MFCD05738885
Formel:
C₁₅H₁₀ClN₃ Molecular Weight: 267,72 g/mol Schmelzpunkt: 138…140 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD05738885 CAS-Nummer: 3842-55-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...