4-Chloro-6,7,8,9-tetrahydro-5H-cyclohepta[4,5]thieno[2,3-d]pyrimidine 96%

Lieferant: Thermo Fisher Scientific
Achtung

H33184.03 H33184.06
H33184.03EA 53.5 EUR
H33184.03 H33184.06
4-Chloro-6,7,8,9-tetrahydro-5H-cyclohepta[4,5]thieno[2,3-d]pyrimidine 96%
4-Chloro-6,7,8,9-tetrahydro-5H-cyclohepta[4,5]thieno[2,3-d]pyrimidine

Purity: 96% CAS No.: 40106-58-9 Molecular Formula: C11H11ClN2S Formula Weight: 238.74 Melting Point: 64-66° MDL No.: MFCD05865130

Formel: C₁₁H₁₁ClN₂S
Molecular Weight: 238.74 g/mol
Schmelzpunkt: 64…66 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD05865130
CAS-Nummer: 40106-58-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR