3-(2,6-Dichlorophenyl)propionic acid 96%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
Synonyme:
3-(2,6-Dichlorophenyl)propanoic acid
H33210.03
H33210.06
H33210.14
H33210.03EA
14.9
EUR
H33210.03
H33210.06
H33210.14
3-(2,6-Dichlorophenyl)propionic acid 96%
3-(2,6-Dichlorophenyl)propionic acid
Purity: 96%
CAS No.: 51656-68-9
Molecular Formula: C9H8Cl2O2
Formula Weight: 219.07
Melting Point: 97-99°
MDL No.: MFCD01310807
Formel:
C₉H₈Cl₂O₂ Schmelzpunkt: 97…99 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD01310807 CAS-Nummer: 51656-68-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...