4-(Trifluormethyl)cyclohexanol (cis/trans-Gemisch) 97%
Lieferant: Thermo Scientific
Achtung
Synonyme: 4-(Trifluoromethyl)cyclohexanol, 4-(Trifluoromethyl)cyclohexan-1-ol
H33317.03 H33317.06
H33317.03EA 78 EUR
H33317.03 H33317.06
4-(Trifluormethyl)cyclohexanol (cis/trans-Gemisch) 97%
4-(Trifluoromethyl)cyclohexanol (cis- and trans- mixture)
Purity: 97% CAS No.: 30129-18-1 Molecular Formula: C7H11F3O Formula Weight: 168.16 Boiling Point: 70-75°/15mm MDL No.: MFCD00102144
Formel: C₇H₁₁F₃O
Molecular Weight: 168,16 g/mol
Siedepunkt: 70…75 °C (15 mmHg)
Lagertemperatur: Raumtemperatur
MDL: MFCD00102144
CAS-Nummer: 30129-18-1