1-(4-Methylpyridin-3-yl)ethanone 96%
Lieferant: Thermo Scientific
|
Gefahr
|
Synonyme:
1-(4-Methyl-3-pyridinyl)-Ethanone
H33380.03
H33380.06
H33380.03EA
59.3
EUR
H33380.03
H33380.06
1-(4-Methylpyridin-3-yl)ethanone 96%
1-(4-Methylpyridin-3-yl)ethanone
Purity: 96%
CAS No.: 51227-30-6
Molecular Formula: C8H9NO
Formula Weight: 135.17
Refractive Index: 1.5295
Boiling Point: 57-58°/1mm
MDL No.: MFCD08447082
Formel:
C₈H₉NO Molecular Weight: 135,17 g/mol Siedepunkt: 57…58 °C (1 mmHg) Lagertemperatur: Raumtemperatur |
MDL:
MFCD08447082 CAS-Nummer: 51227-30-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...