4-n-Nonylaniline 98%

Lieferant: Thermo Fisher Scientific
Gefahr

H33414.06 H33414.14 H33414.03
H33414.06EA 80.3 EUR
H33414.06 H33414.14 H33414.03
4-n-Nonylaniline 98%
4-n-Nonylaniline

Purity: 98% CAS No.: 37529-29-6 Molecular Formula: C15H25N Formula Weight: 219.37 Flash Point: 110°(230°F) Boiling Point: 144/2mm MDL No.: MFCD00038370

Formel: C₁₅H₂₅N
Molecular Weight: 219.37 g/mol
Siedepunkt: 144 °C (2 mmHg)
Flammpunkt: >110 °C (230 °F)
Lagertemperatur: Raumtemperatur
MDL: MFCD00038370
CAS-Nummer: 37529-29-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR