3-Amino-5-phenyl-1,2,4-thiadiazole 96%

Lieferant: Thermo Scientific
Gefahr

H33552.MD H33552.03 H33552.06
H33552.MDEA 35.7 EUR
H33552.MD H33552.03 H33552.06
3-Amino-5-phenyl-1,2,4-thiadiazole 96%
3-Amino-5-phenyl-1,2,4-thiadiazole

Purity: 96% CAS No.: 27182-54-3 Molecular Formula: C8H7N3S Formula Weight: 177.23 Melting Point: 136-137° MDL No.: MFCD00159799

Formel: C₈H₇N₃S
Molecular Weight: 177,23 g/mol
Schmelzpunkt: 136…137 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00159799
CAS-Nummer: 27182-54-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR