2,3-Dihydro-1-benzofuran-5-sulfonylchlorid 97%

Lieferant: Thermo Scientific
Gefahr

H33579.MD H33579.06
H33579.MDEA 29.7 EUR
H33579.MD H33579.06
2,3-Dihydro-1-benzofuran-5-sulfonylchlorid 97%
2,3-Dihydro-1-benzofuran-5-sulphonoylchloride

Purity: 97% CAS No.: 115010-11-2 Molecular Formula: C8H7ClO3S Formula Weight: 218.66 Storage Temperature: 2°C to 8°C Melting Point: 83-85° UN No.: UN3261 MDL No.: MFCD02677703

Formel: C₈H₇ClO₃S
Molecular Weight: 218,66 g/mol
Schmelzpunkt: 83…85 °C
MDL: MFCD02677703
CAS-Nummer: 115010-11-2
ADR: 8,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR