2,3,5-Tri-O-benzyl-D-ribofuranose 98%

Lieferant: Thermo Scientific
Achtung

Synonyme: (2R,3R,4R)-2,3,5-Tris(benzyloxy)-4-hydroxypentanal

H33605.03 H33605.06
H33605.03EA 80.6 EUR
H33605.03 H33605.06
2,3,5-Tri-O-benzyl-D-ribofuranose 98%
2,3,5-Tri-O-benzyl-D-ribofuranose

Purity: 98% CAS No.: 54623-25-5 Molecular Formula: C26H28O5 Formula Weight: 420.50 MDL No.: MFCD03425640

Formel: C₂₆H₂₈O₅
Molecular Weight: 420,50 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD03425640
CAS-Nummer: 54623-25-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR