4-(Thiophen-2-yl)benzoic acid 96%

Lieferant: Thermo Scientific
Gefahr

H33642.03 H33642.06
H33642.03EA 59.8 EUR
H33642.03 H33642.06
4-(Thiophen-2-yl)benzoic acid 96%
4-(Thiophen-2-yl)benzoic acid

Purity: 96% CAS No.: 29886-62-2 Molecular Formula: C11H8O2S Formula Weight: 204.25 Melting Point: 243-245° MDL No.: MFCD00690742

Formel: C₁₁H₈O₂S
Molecular Weight: 204,25 g/mol
Schmelzpunkt: 243…245 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00690742
CAS-Nummer: 29886-62-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR