Ethyl 2-amino-5-phenylthiophene-3-carboxylate 97%

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: Ethyl 2-amino-5-phenylthiophene-3-carboxylate

H33823.03 H33823.06
H33823.03EA 92 EUR
H33823.03 H33823.06
Ethyl 2-amino-5-phenylthiophene-3-carboxylate 97%
Ethyl-2-amino-5-phenylthiophene-3-carboxylate

Purity: 97% CAS No.: 4815-34-3 Molecular Formula: C13H13NO2S Formula Weight: 247.32 Melting Point: 121-123° MDL No.: MFCD01829801

Formel: C₁₃H₁₃NO₂S
Schmelzpunkt: 121…123 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD01829801
CAS-Nummer: 4815-34-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR