3-[3-(Benzyloxy)phenyl]propionic acid 96%
Lieferant: Thermo Scientific
H33899.03
H33899.06
H33899.03EA
82.1
EUR
H33899.03
H33899.06
3-[3-(Benzyloxy)phenyl]propionic acid 96%
3-[3-(Benzyloxy)phenyl]propionic acid
Purity: 96%
CAS No.: 57668-34-5
Molecular Formula: C16H16O3
Formula Weight: 256.30
Melting Point: 83-85°
MDL No.: MFCD09258905
Formel:
C₁₆H₁₆O₃ Molecular Weight: 256,30 g/mol Schmelzpunkt: 83…85 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD09258905 CAS-Nummer: 57668-34-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...