7-Benzyloxy-6-methoxy-3H-quinazolin-4-one 96%
Lieferant: Thermo Fisher Scientific
Synonyme: 7-Benzyloxy-4-hydroxy-6-methoxyquinazoline, 7-Benzyloxy-6-methoxy-4-quinazolinol, 7-(Benzyloxy)-6-methoxyquinazolin-4(3H)-one
H34131.03 H34131.06
H34131.03EA 43.5 EUR
H34131.03 H34131.06
7-Benzyloxy-6-methoxy-3H-quinazolin-4-one 96%
7-Benzyloxy-6-methoxy-3H-quinazolin-4-one
Purity: 96% CAS No.: 179688-01-8 Molecular Formula: C16H16N2O3 Formula Weight: 284.31 MDL No.: MFCD04115119
Formel: C₁₆H₁₄N₂O₃
Molecular Weight: 282.30 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD04115119
CAS-Nummer: 179688-01-8