2-Ethoxybenzenesulphonyl chloride 96%

Lieferant: Thermo Scientific
Gefahr

H34325.03 H34325.06
H34325.03EA 70 EUR
H34325.03 H34325.06
2-Ethoxybenzenesulphonyl chloride 96%
2-Ethoxybenzenesulphonyl chloride

Purity: 96% CAS No.: 68800-33-9 Molecular Formula: C8H9ClO3S Formula Weight: 220.67 Melting Point: 60-64° UN No.: UN3261 MDL No.: MFCD08706420

Formel: C₈H₉ClO₃S
Molecular Weight: 220,67 g/mol
Schmelzpunkt: 60…64 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD08706420
CAS-Nummer: 68800-33-9
ADR: 8,II

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR