2-Chloro-1H-indole-3-carbaldehyde 97%

Lieferant: Thermo Scientific
Achtung

H34381.03 H34381.09
H34381.03EA 54.4 EUR
H34381.03 H34381.09
2-Chloro-1H-indole-3-carbaldehyde 97%
2-Chloro-1H-indole-3-carbaldehyde

Purity: 97% CAS No.: 5059-30-3 Molecular Formula: C9H6ClNO Formula Weight: 179.61 Melting Point: ca 212° dec. MDL No.: MFCD00111620

Formel: C₉H₆ClNO
Molecular Weight: 179,61 g/mol
Schmelzpunkt: ∼ 212 °C (Zers.)
Lagertemperatur: Raumtemperatur
MDL: MFCD00111620
CAS-Nummer: 5059-30-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR