Methylblau
Lieferant: Thermo Fisher Scientific
Achtung
Synonyme: Baumwollblau, Acid Blue 93, Anilinblau WS, Wasserblau 6B, Aniline blue water soluble, Anilinblau wasserlöslich, Aniline blue WS, Dinatrium-[[4-[bis[4-[(sulfonatophenyl)amino]phenyl]methylen]cyclohexa-2,5-dien-1-yliden]amino]benzolsulfonat
H37721.14 H37721.22
H37721.14EA 27.3 EUR
H37721.14 H37721.22
Methylblau
Methylblau
CAS No.: 28983-56-4 Molecular Formula: C37H27N3Na2O9S3 Formula Weight: 799.80 Physical Form: Powder Appearance: Blue Odour: Odourless MDL No.: MFCD00058509
Formel: C₃₇H₂₇N₃Na₂O₉S₃
Molecular Weight: 799.81 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00058509
CAS-Nummer: 28983-56-4
EINECS: 249-352-9
Merck Index: 13,06055