2-Ethoxypyridine-3-carboxaldehyde 97%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
Synonyme:
2-Ethoxypyridine-3-carbaldehyde, 2-Ethoxynicotinaldehyde
H50088.03
H50088.06
H50088.03EA
105
EUR
H50088.03
H50088.06
2-Ethoxypyridine-3-carboxaldehyde 97%
2-Ethoxypyridine-3-carboxaldehyde
Purity: 97%
CAS No.: 885278-07-9
Molecular Formula: C8H9NO2
Formula Weight: 151.17
MDL No.: MFCD06804559
Formel:
C₈H₉NO₂ Molecular Weight: 151.17 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD06804559 CAS-Nummer: 885278-07-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...