2-Isopropoxypyridine-3-carboxaldehyde 98%
Lieferant: Thermo Scientific
|
Achtung
|
Synonyme:
2-Isopropoxynicotinaldehyde
H50089.03
H50089.06
H50089.03EA
105
EUR
H50089.03
H50089.06
2-Isopropoxypyridine-3-carboxaldehyde 98%
2-Isopropoxypyridine-3-carboxaldehyde
Purity: 98%
CAS No.: 885278-10-4
Molecular Formula: C9H11NO2
Formula Weight: 165.19
MDL No.: MFCD06804560
Formel:
C₉H₁₁NO₂ Molecular Weight: 165,19 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD06804560 CAS-Nummer: 885278-10-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...