5-Benzyloxy-6-azaindole-3-carboxaldehyde 96%
Lieferant: Thermo Scientific
|
Achtung
|
H50173.03
H50173.MD
H50173.03EA
154
EUR
H50173.03
H50173.MD
5-Benzyloxy-6-azaindole-3-carboxaldehyde 96%
5-Benzyloxy-6-azaindole-3-carboxaldehyde
Purity: 96%
CAS No.: 56795-92-7
Molecular Formula: C15H12N2O2
Formula Weight: 252.28
MDL No.: MFCD00191784
Formel:
C₁₅H₁₂N₂O₂ Molecular Weight: 252,28 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00191784 CAS-Nummer: 56795-92-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...