Boc-Cys(Et)-OH

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: (R)-2-((tert-Butoxycarbonyl)amino)-3-(ethylthio)propanoic acid

H50216.03 H50216.06
H50216.03EA 86 EUR
H50216.03 H50216.06
Boc-Cys(Et)-OH
Boc-Cys(Et)-OH

CAS No.: 16947-82-3 Molecular Formula: C10H19NO4S Formula Weight: 249.33 MDL No.: MFCD00076919

Formel: C₁₀H₁₉NO₄S
Molecular Weight: 249.325 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00076919
CAS-Nummer: 16947-82-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR