N-Boc-S-methyl-L-cysteine 96%

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: Boc-Cys(Me)-OH

H50217.03 H50217.06
H50217.03EA 82.5 EUR
H50217.03 H50217.06
N-Boc-S-methyl-L-cysteine 96%
N-Boc-S-methyl-L-cysteine

Purity: 96% CAS No.: 16947-80-1 Molecular Formula: C9H17NO4S Formula Weight: 235.30 MDL No.: MFCD00076923

Formel: C₉H₁₇NO₄S
Molecular Weight: 235.298 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00076923
CAS-Nummer: 16947-80-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR