4-(Methylsulphonyl)benzenesulphonyl chloride 95%
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
Synonyme:
4-(Methylsulphonyl)benzene-1-sulphonyl chloride
H50264.03
H50264.06
H50264.03EA
61.5
EUR
H50264.03
H50264.06
4-(Methylsulphonyl)benzenesulphonyl chloride 95%
4-(Methylsulphonyl)benzenesulphonyl chloride
Purity: 95%
CAS No.: 82964-91-8
Molecular Formula: C7H7ClO4S2
Formula Weight: 254.71
Melting Point: 158-163°
UN No.: UN3261
MDL No.: MFCD00216486
Formel:
C₇H₇ClO₄S₂ Molecular Weight: 254.699 g/mol Schmelzpunkt: 158…163 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00216486 CAS-Nummer: 82964-91-8 ADR: 8 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...