1-Methyl-1H-pyrazole-3-carboxylic acid 96%
Lieferant: Thermo Scientific
Achtung
Synonyme: 1-Methylpyrazole-3-carboxylic acid
H50317.03 H50317.MD
H50317.03EA 143 EUR
H50317.03 H50317.MD
1-Methyl-1H-pyrazole-3-carboxylic acid 96%
1-Methyl-1H-pyrazole-3-carboxylic acid
Purity: 96% CAS No.: 25016-20-0 Molecular Formula: C5H6N2O2 Formula Weight: 126.11 Odour: Odourless Melting Point: 146-156° MDL No.: MFCD00464254
Formel: C₅H₆N₂O₂
Molecular Weight: 126,12 g/mol
Schmelzpunkt: 146…156 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00464254
CAS-Nummer: 25016-20-0