N-Benzhydryl-N-methylamine 98%

Lieferant: Thermo Scientific
Achtung

Synonyme: N-Methylbenzhydrylamine

H50327.03 H50327.06
H50327.03EA 41.3 EUR
H50327.03 H50327.06
N-Benzhydryl-N-methylamine 98%
N-Benzhydryl-N-methylamine

Purity: 98% CAS No.: 14683-47-7 Molecular Formula: C14H15N Formula Weight: 197.28 Melting Point: 38-42° Beilstein Reference: 2096409 MDL No.: MFCD00467853

Formel: C₁₄H₁₅N
Molecular Weight: 197,28 g/mol
Schmelzpunkt: 38…42 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00467853
CAS-Nummer: 14683-47-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR