2-Pyrazinecarbonyl chloride ≥90%

Lieferant: Thermo Scientific
Gefahr

Synonyme: Pyrazine-2-carbonyl chloride

H50418.03 H50418.06
H50418.03EA 156 EUR
H50418.03 H50418.06
2-Pyrazinecarbonyl chloride ≥90%
2-Pyrazinecarbonyl chloride

Purity: 90+% CAS No.: 19847-10-0 Molecular Formula: C5H3ClN2O Formula Weight: 142.55 UN No.: UN3261 MDL No.: MFCD00792886

Formel: C₅H₃ClN₂O
Lagertemperatur: Raumtemperatur
MDL: MFCD00792886
CAS-Nummer: 19847-10-0
EINECS: 243-367-4
ADR: 8,II

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR