5-(4-Methoxyphenyl)-2-thiophenecarbaldehyde 99%

Lieferant: Thermo Fisher Scientific
Achtung

H50483.03 H50483.06
H50483.03EA 83.3 EUR
H50483.03 H50483.06
5-(4-Methoxyphenyl)-2-thiophenecarbaldehyde 99%
5-(4-Methoxyphenyl)-2-thiophenecarbaldehyde

Purity: 99% CAS No.: 38401-67-1 Molecular Formula: C12H10O2S Formula Weight: 218.28 MDL No.: MFCD00663639

Formel: C₁₂H₁₀O₂S
Molecular Weight: 218.28 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00663639
CAS-Nummer: 38401-67-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR