N-(4-Bromo-2,6-dimethylphenyl)-N,N-dimethylamine 98%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
H50584.03
H50584.06
H50584.03EA
96
EUR
H50584.03
H50584.06
N-(4-Bromo-2,6-dimethylphenyl)-N,N-dimethylamine 98%
N-(4-Bromo-2,6-dimethylphenyl)-N,N-dimethylamine
Purity: 98%
CAS No.: 50638-54-5
Molecular Formula: C10H14BrN
Formula Weight: 228.13
MDL No.: MFCD00100113
Formel:
C₁₀H₁₄BrN Molecular Weight: 228.14 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00100113 CAS-Nummer: 50638-54-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...