2-[4-(1,3,4-Oxadiazol-2-yl)phenoxy]acetamidoxime 97%

Lieferant: Thermo Scientific
Achtung

H50673.03 H50673.MD
H50673.03EA 164 EUR
H50673.03 H50673.MD
2-[4-(1,3,4-Oxadiazol-2-yl)phenoxy]acetamidoxime 97%
2-[4-(1,3,4-Oxadiazol-2-yl)phenoxy]acetamidoxime

Purity: 97% CAS No.: 258521-20-9 Molecular Formula: C10H10N4O3 Formula Weight: 234.22 MDL No.: MFCD01764763

Formel: C₁₀H₁₀N₄O₃
Molecular Weight: 234,22 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD01764763
CAS-Nummer: 258521-20-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR