2-Hydrazinochinolin 97%

Lieferant: Thermo Scientific
Achtung

Synonyme: 2-Quinolylhydrazine, 2-Hydrazinylquinoline

H50700.03 H50700.MD
H50700.03EA 199 EUR
H50700.03 H50700.MD
2-Hydrazinochinolin 97%
2-Hydrazinoquinoline

Purity: 97% CAS No.: 15793-77-8 Molecular Formula: C9H9N3 Formula Weight: 159.19 Melting Point: 141-143° MDL No.: MFCD00041860

Formel: C₉H₉N₃
Molecular Weight: 159,19 g/mol
Schmelzpunkt: 141…143 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00041860
CAS-Nummer: 15793-77-8
EINECS: 239-899-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR