3,4-Methylenedioxybenzhydrazide 97%

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: Benzo[d][1,3]dioxole-5-carbohydrazide

H50713.03 H50713.06
H50713.03EA 43.9 EUR
H50713.03 H50713.06
3,4-Methylenedioxybenzhydrazide 97%
3,4-Methylenedioxybenzhydrazide

Purity: 97% CAS No.: 22026-39-7 Molecular Formula: C8H8N2O3 Formula Weight: 180.16 Melting Point: 176-178° MDL No.: MFCD00060505

Formel: C₈H₈N₂O₃
Molecular Weight: 180.16 g/mol
Schmelzpunkt: 176…178 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00060505
CAS-Nummer: 22026-39-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR