4-Phenyltetrahydropyran-4-carboxaldehyde 95%

Lieferant: Thermo Scientific
Achtung

Synonyme: 4-Phenyltetrahydro-2H-pyran-4-carbaldehyde

H50816.03 H50816.MD
H50816.03EA 293 EUR
H50816.03 H50816.MD
4-Phenyltetrahydropyran-4-carboxaldehyde 95%
4-Phenyltetrahydropyran-4-carboxaldehyde

Purity: 95% CAS No.: 66109-88-4 Molecular Formula: C12H14O2 Formula Weight: 190.24 MDL No.: MFCD08061006

Formel: C₁₂H₁₄O₂
Molecular Weight: 190,24 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD08061006
CAS-Nummer: 66109-88-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR