N-Hydroxypropionamidine 97%

Lieferant: Thermo Scientific
Achtung

Synonyme: Propionamidoxime

H50889.03 H50889.MD
H50889.03EA 152 EUR
H50889.03 H50889.MD
N-Hydroxypropionamidine 97%
N-Hydroxypropionamidine

Purity: 97% CAS No.: 29335-36-2 Molecular Formula: C3H8N2O Formula Weight: 88.11 MDL No.: MFCD03426272

Formel: C₃H₈N₂O
Molecular Weight: 88,11 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD03426272
CAS-Nummer: 29335-36-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR