6-Methoxy-1,3-benzothiazol-2-carbonitril 99%

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: 6-Methoxy-2-benzothiazolecarbonitrile, 6-Methoxybenzo[d]thiazole-2-carbonitrile

H51082.03 H51082.06
H51082.03EA 119 EUR
H51082.03 H51082.06
6-Methoxy-1,3-benzothiazol-2-carbonitril 99%
2-Cyano-6-methoxybenzothiazole

Purity: 99% CAS No.: 943-03-3 Molecular Formula: C9H6N2OS Formula Weight: 190.22 MDL No.: MFCD00010537

Formel: C₉H₆N₂OS
Lagertemperatur: Raumtemperatur
MDL: MFCD00010537
CAS-Nummer: 943-03-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR