Uracil-5-carboxaldehyde 97%
Lieferant: Thermo Scientific
Synonyme:
2,4-Dioxo-1,2,3,4-tetrahydropyrimidine-5-carbaldehyde, 5-Formyluracil
H51098.MD
H51098.03
H51098.MDEA
34.8
EUR
H51098.MD
H51098.03
Uracil-5-carboxaldehyde 97%
Uracil-5-carboxaldehyde
Purity: 97%
CAS No.: 1195-08-0
Molecular Formula: C5H4N2O3
Formula Weight: 140.10
MDL No.: MFCD00192185
Formel:
C₅H₄N₂O₃ Molecular Weight: 140,10 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00192185 CAS-Nummer: 1195-08-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...