4-Ethynylbenzeneboronic acid pinacol ester 95%
Lieferant: Thermo Scientific
Synonyme:
2-(4-Ethynyl-phenyl)-4,4,5,5-tetramethyl-[1,3,2]dioxaborolane, 2-(4-Ethynyl-phenyl)-4,4,5,5-tetramethyl-[1,3,2]-dioxaborolane
H51745.03
H51745.06
H51745.03EA
104
EUR
H51745.03
H51745.06
4-Ethynylbenzeneboronic acid pinacol ester 95%
4-Ethynylbenzeneboronic acid pinacol ester
Purity: 95%
CAS No.: 1034287-04-1
Molecular Formula: C14H17BO2
Formula Weight: 228.10
MDL No.: MFCD16294504
Formel:
C₁₄H₁₇BO₂ Molecular Weight: 228,10 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD16294504 CAS-Nummer: 1034287-04-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...