2-(4-Pyridyl)benzimidazole 97%

Lieferant: Thermo Scientific
Achtung

Synonyme: 2-(Pyridin-4-yl)-1H-benzo[d]imidazole

H51778.06 H51778.14
H51778.06EA 67.7 EUR
H51778.06 H51778.14
2-(4-Pyridyl)benzimidazole 97%
2-(4-Pyridyl)benzimidazole

Purity: 97% CAS No.: 2208-59-5 Molecular Formula: C12H9N3 Formula Weight: 195.22 Odour: Odourless Melting Point: 216-221° MDL No.: MFCD00453948

Formel: C₁₂H₉N₃
Molecular Weight: 195,23 g/mol
Schmelzpunkt: 216…221 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00453948
CAS-Nummer: 2208-59-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR