2-Pyridin-3-yl-1H-benzimidazole 97%
Lieferant: Thermo Scientific
|
Achtung
|
Synonyme:
2-(Pyridin-3-yl)-1H-benzo[d]imidazole
H51785.06
H51785.14
H51785.06EA
62.2
EUR
H51785.06
H51785.14
2-Pyridin-3-yl-1H-benzimidazole 97%
2-Pyridin-3-yl-1H-benzimidazole
Purity: 97%
CAS No.: 1137-67-3
Molecular Formula: C12H9N3
Formula Weight: 195.22
Odour: Odourless
Melting Point: 254-260°
MDL No.: MFCD00022665
Formel:
C₁₂H₉N₃ Molecular Weight: 195,23 g/mol Schmelzpunkt: 254…260 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00022665 CAS-Nummer: 1137-67-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...