Benzene-1,4-dithiocarboxamide 97%
Lieferant: Thermo Scientific
|
Gefahr
|
H51857.03
H51857.06
H51857.03EA
78.2
EUR
H51857.03
H51857.06
Benzene-1,4-dithiocarboxamide 97%
Benzene-1,4-dithiocarboxamide
Purity: 97%
CAS No.: 13363-51-4
Molecular Formula: C8H8N2S2
Formula Weight: 196.28
UN No.: UN2811
MDL No.: MFCD00046866
Formel:
C₈H₈N₂S₂ Molecular Weight: 196,28 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00046866 CAS-Nummer: 13363-51-4 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...