Benzo[b]thiophene-3-boronic acid pinacol ester 95%
Lieferant: Thermo Scientific
Synonyme:
2-(Benzo[b]thiophen-3-yl)-4,4,5,5-tetramethyl-[1,3,2]dioxaborolane, 2-(Benzo[b]thiophen-3-yl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
H51876.MD
H51876.03
H51876.MDEA
53.2
EUR
H51876.MD
H51876.03
Benzo[b]thiophene-3-boronic acid pinacol ester 95%
Benzo[b]thiophene-3-boronic acid pinacol ester
Purity: 95%
CAS No.: 171364-86-6
Molecular Formula: C14H17BO2S
Formula Weight: 260.16
MDL No.: MFCD11110552
Formel:
C₁₄H₁₇BO₂S Molecular Weight: 260,16 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD11110552 CAS-Nummer: 171364-86-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...