Fluorene-2-boronic acid pinacol ester 95%

Lieferant: Thermo Fisher Scientific

Synonyme: 9H-Fluorene-2-boronic acid Pinacol ester, 2-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolane-2-yl)-9H-fluorene, 2-(9H-Fluoren-2-yl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 2-(9H-Fluoren-2-yl)-4,4,5,5-tetramethyl-[1,3,2]dioxaborolane

H51947.03 H51947.06
H51947.03EA 35.3 EUR
H51947.03 H51947.06
Fluorene-2-boronic acid pinacol ester 95%
Fluorene-2-boronic acid pinacol ester

Purity: 95% Molecular Formula: C19H21BO2 Formula Weight: 292.18 MDL No.: MFCD16294548

Formel: C₁₉H₂₁BO₂
Molecular Weight: 292.18 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD16294548
CAS-Nummer: 922706-40-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR